missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ LOX Polyclonal Antibody
GREENER_CHOICE

Product Code. 11561842
Change view
Click to view available options
Quantity:
100 μL
Unit Size:
100µL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11561842 100 μL 100µL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 11561842 Supplier Invitrogen™ Supplier No. PA116989

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

This is a highly glycosylated protein and can exhibit bands at ~32 kDa, ~50 kDa and ~58 kDa representing the mature (secreted) form, the pro form and the glycosylated forms, respectively. Suggested positive control: MC3T3-E1 lysate.

Lysyl oxidase (LOX), a copper-containing amine oxidase, belongs to a heterogeneous family of enzymes that oxidize primary amine substrates to reactive aldehydes. It plays a vital role in the formation and repair of the extra cellular matrix. As well, LOX is a multifunctional enzyme having diverse biological functions such as developmental regulation, tumor suppression, cell motility and cellular senescence. The secreted form of LOX is responsible for the invasive properties of hypoxic human cancer cells. Thus, it is essential for hypoxia-induced metastasis and is a good therapeutic target for preventing and treating metastases.
TRUSTED_SUSTAINABILITY

Specifications

Antigen LOX
Applications Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry, Western Blot
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 0.05% sodium azide
Gene LOX
Gene Accession No. P16636, P28300, P28301, P45845
Gene Alias AI893619; cross-linking enzyme; H-rev142; LOX; Lysyl oxidase; Lysyl oxidase (an H-rev gene with its expression down-regulated in HRAS-transformed rat 208F fibroblasts); MGC105112; protein-lysine 6-oxidase; Protein-lysine 6-oxidase, long form; Protein-lysine 6-oxidase, short form; Ras excision protein; ras recision; rrg; Rrg1; TSC-160
Gene Symbols LOX
Host Species Rabbit
Immunogen A synthetic peptide made to an internal region of mouse LOX propeptide (residues 78-115).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 100525278, 16948, 24914, 4015
Target Species Human, Mouse, Rat, Pig, Rat
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.