missing translation for 'onlineSavingsMsg'
Learn More
Learn More
LOX Antibody, DyLight 550, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NB100-2530R
This item is not returnable.
View return policy
Description
LOX Polyclonal specifically detects LOX in Human, Mouse, Rat, Porcine, Bovine samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin.
Specifications
| LOX | |
| Polyclonal | |
| Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin | |
| P28300 | |
| LOX | |
| A cocktail of two synthetic peptides; one made to a region of the human LOX protein within residues 300-350 (NB100-2528) and one within residues 200-300 (NB100-2527). [Swiss-Prot P28300] | |
| 0.05 mL | |
| Cancer, Cytoskeleton Markers, Endothelial Cell Markers, Extracellular Matrix, Hypoxia, Signal Transduction | |
| 4015 | |
| Human, Mouse, Rat, Pig, Bovine | |
| Purified |
| Western Blot, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin) | |
| DyLight 550 | |
| 50mM Sodium Borate with 0.05% Sodium Azide | |
| lysyl oxidaseEC 1.4.3.13, MGC105112, protein-lysine 6-oxidase | |
| Rabbit | |
| Immunogen affinity purified | |
| RUO | |
| Primary | |
| Human, mouse, rat, cow and pig. 92% sequence identity with chicken, Zebrafish, and Xenopus proteins. | |
| Store at 4C in the dark. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction