Learn More
Invitrogen™ LOX-1 Polyclonal Antibody
Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA595063
Description
Reconstitute with 0.2 mL of distilled water to yield a concentration of 500 μg/mL. Positive Control - WB: human prostatic carcinoma tissue (PCT), human cervical carcinoma tissue (CCAT), human gallbladder carcinoma tissue (GBCT), human intrahepatic cholangiocarcinom paracancerous tissue (ICCP). Store at -20°C for one year from date of receipt. After reconstitution, at 4°C for one month. It can also be aliquotted and stored frozen at -20°C for six months. Avoid repeated freeze-thaw cycles.
LOX-1 (lectin-like oxidized LDL receptor -1) is a 40-50 kDa type II membrane glycoprotein class E scavenger receptor expressed by endothelial cells, macrophages, activated platelets and smooth muscle cells. LOX-1 binds to a wide range of ligands, including oxidized low-density lipoprotein (oxLDL), hypochlorite modified high-density lipoprotein (HDL), aged /apoptotic cells, activated platelets and bacteria, reflecting its versatile physiological functions.
Specifications
| LOX-1 | |
| Polyclonal | |
| Unconjugated | |
| OLR1 | |
| CLEC8A; C-type lectin domain family 8 member A; hLOX 1; hLOX1; hLOX-1; lectin-like oxidized LDL receptor; lectin-like oxidized LDL receptor 1; lectin-like oxidized low-density lipoprotein receptor; Lectin-like oxidized low-density lipoprotein receptor-1; lectin-like oxLDL receptor 1; lectin-type oxidized LDL receptor 1; LOX 1; LOX1; LOX-1; LOXIN; Oldlr1; Oldr1; OLR1; ox LDL receptor 1; oxidised low density lipoprotein (lectin-like) receptor 1; oxidized low density lipoprotein (lectin-like) receptor 1; oxidized low density lipoprotein receptor 1; oxidized low-density lipoprotein receptor 1; Oxidized low-density lipoprotein receptor 1, soluble form; ox-LDL receptor 1; Scare1; scavenger receptor class E, member 1; sLOX 1; SLOX1; soluble LOX 1; SR-EI | |
| Rabbit | |
| Affinity chromatography | |
| RUO | |
| 4973 | |
| -20°C | |
| Lyophilized |
| Western Blot | |
| 500 μg/mL | |
| PBS with 5mg BSA and 0.05mg sodium azide, 0.05mg thimerosal | |
| P78380 | |
| OLR1 | |
| A synthetic peptide corresponding to a sequence at the N-terminus of human LOX 1 (1-16aa MTFDDLKIQTVKDQPD). | |
| 100 μg | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.