missing translation for 'onlineSavingsMsg'
Learn More

Buerkle™ Lowering Cable for Buerkle™ Dipping Bottles

Product Code. 11882663 Shop All Bürkle Products
Change view
Click to view available options
Length (Metric):
10 m
25 m
50 m
100 m
Material:
Copper
Polypropylene
Stainless Steel
Stainless Steel (V2A)
Diameter (Metric) Outer:
1.25 mm
2 mm
2.3 mm
4.5 mm
Unit Size:
Each
15 product options available for selection
Product selection table with 15 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Length (Metric) Material Diameter (Metric) Outer unitSize
11882663 10 m Copper 4.5 mm Each
11892663 25 m Copper 4.5 mm Each
11802673 50 m Copper 4.5 mm Each
10066361 50 m Polypropylene 2 mm Each
10157871 25 m Polypropylene 2 mm Each
10279771 10 m Polypropylene 2 mm Each
10107781 10 m Stainless Steel (V2A) 1.25 mm Each
10498421 25 m Stainless Steel (V2A) 1.25 mm Each
10084800 10 m Stainless Steel (V2A) 2 mm Each
10779972 25 m Stainless Steel (V2A) 2 mm Each
10408431 50 m Stainless Steel (V2A) 1.25 mm Each
10381132 50 m Stainless Steel (V2A) 2 mm Each
17270574 25 m - - Each
10579902 10 m Stainless Steel 2.3 mm Each
17280574 100 m - - Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 15 options available.
15 options
This item is not returnable. View return policy
Product Code. 11882663 Supplier Buerkle™ Supplier No. 53090410
Only null left
Add to basket
Add to basket
This item is not returnable. View return policy

Buerkle™ Lowering Cable for dipping bottles and immersion cylinders, matching manually-operated drum reels made of various materials.

  • Good chemical resistance
  • For dipping bottles and immersion cylinders, matching manually-operated drum reels made of various materials
  • Tensile load max. up to 10kg
TRUSTED_SUSTAINABILITY

Specifications

Product Type Lowering Cable
Material Copper
For Use With (Application) Dipping Samplers and Immersion Cylinders
Diameter (Metric) Outer 4.5 mm
Length (Metric) 10 m
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.