missing translation for 'onlineSavingsMsg'
Learn More

Buerkle™ Lowering Cable for Buerkle™ Dipping Bottles

Buerkle™ Lowering Cable for dipping bottles and immersion cylinders, matching manually-operated drum reels made of various materials.

Brand:  Buerkle™ 5309-0202

Code : 03

Additional Details : Weight : 0.07450kg

 View more versions of this product

Product Code. 10157871

  • £27.80 / Each

Estimated Shipment: 09-02-2022
Add to basket

Description

Description

  • Good chemical resistance
  • For dipping bottles and immersion cylinders, matching manually-operated drum reels made of various materials
  • Tensile load max. up to 10kg
Specifications

Specifications

Polypropylene
Lowering cable
25 m
Red
Dipping Bottles and Immersion Cylinders, Manually Operated Drum Reels
2 mm
Videos
SDS
Documents
Certificates
Special Offers

Special Offers