Learn More
Lovibond™ Replacement Reagents for CHECKIT™ Comparator: Phosphate No. 2 LR Tablets
Lovibond Replacement Reagents for CHECKIT Comparator: Phosphate No. 2 LR Tablets, Range: 0.05 to 4mg/L, White, Composition: Boric Acid 70 to 80%, Disodium Disulphite 10 to 20%, Ammonium Heptamolybdate Tetrahydrate 5 to 10%, non-hazardous components 2.5%, Format: Tablet
Brand: Lovibond™ 513050BT
Code : NEW
Additional Details : Weight : 0.27000kg
Description
Lovibond Replacement Reagents for CHECKIT Comparator: Phosphate No. 2 LR Tablets, Range: 0.05 to 4mg/L, White, Composition: Boric Acid 70 to 80%, Disodium Disulphite 10 to 20%, Ammonium Heptamolybdate Tetrahydrate 5 to 10%, non-hazardous components 2.5%, Format: Tablet
Specifications
White | |
4.72 | |
Low range phosphate AC test kit, phosphate, ortho LR reagent set, water analysis | |
0.05 to 4mg/L | |
Phosphate No. 2 LR Tablet |
Boric Acid 70 to 80%, Disodium Disulphite 10 to 20%, Ammonium Heptamolybdate Tetrahydrate 5 to 10%, non-hazardous components 2.5% | |
Tablet | |
MC500 colorimeters and SP600 spectrophotometers | |
Phosphomolybdic Acid/Ascorbic Acid |
Safety and Handling
- phosphate No. 2 LR
Signal Word
- Danger
Hazard Category
- Serious eye damage/eye irritation Category 1
Hazard Statement
- H318-Causes serious eye damage.
- EUH031-Contact with acids liberates toxic gas.
Precautionary Statement
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P313-Get medical advice/attention.
Supplemental information
- MIXTURE LIST-Contains : disodium disulphate