Learn More
Lovibond™ Replacement Reagent for CHECKIT™ Comparator
Lovibond™ Replacement Reagent for CHECKIT™ Comparator is manufactured under tightly controlled conditions to maintain the highest quality standards.
Brand: Lovibond™ 461181
Code : NEW
Additional Details : CAS Number : 7697-37-2 Weight : 1.05000kg Transport : UN number : 3264 Chem class : 8 Pack group : III
Description
Lovibond™ Replacement Reagent for CHECKIT™ Comparator is manufactured under tightly controlled conditions to maintain the highest quality standards.
- Highest quality production standards
- Consistent and reliable reagents are provided in liquid and tablet formats
- Maximum accuracy
Specifications
Yellow | |
0.1 | |
Water analysis | |
100 mL | |
20°C |
Nitric Acid 2.5 to 5% | |
Liquid | |
Colorimeter and spectrophotometer | |
0 to 0.5mg/L | |
DEHA Test Solution |
Safety and Handling
- DEHA reagent solution
Signal Word
- Danger
Hazard Category
- Substances and mixtures corrosive to metals Category 1
- Serious eye damage/eye irritation Category 1
- Skin corrosion/irritation Category 2
Hazard Statement
- H290-May be corrosive to metals.
- H318-Causes serious eye damage.
- H315-Causes skin irritation.
Precautionary Statement
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P302+P352-IF ON SKIN: Wash with plenty of water/.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P310-Immediately call a POISON CENTER/doctor/.
- P390-Absorb spillage to prevent material damage.
Supplemental information
- MIXTURE LIST-Contains : nitric acid