missing translation for 'onlineSavingsMsg'
Learn More

Lovibond™ Replacement Reagents for CHECKIT™ Comparator: Copper/Zinc LR Tablets

Product Code. 15391765
Click to view available options
Range:
0.02 to 1, 0 to 1mg/L
Unit Size:
Pack of 100
This item is not returnable. View return policy

Product Code. 15391765

Brand: Lovibond™ 512620BT

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Lovibond Replacement Reagents for CHECKIT Comparator: Copper/Zinc LR Tablets, Copper/Zinc Tablet, Range: 0.02 to 1, 0 to 1mg/L, Violet, Composition: 2.5 to 5% Boric acid, 90 to 95% Non-hazardous components, 2.5 to 5% Disodium Tetraborate, anhydrous, Format: Tablet

Specifications

Color Violet
Composition 2.5 to 5% Boric acid, 90 to 95% Non-hazardous components, 2.5 to 5% Disodium Tetraborate, anhydrous
pH Range 8.8
Format Tablet
For Use With (Application) Water analysis
For Use With (Equipment) Colorimeter and spectrophotometer
Range 0.02 to 1, 0 to 1mg/L
Test Method Zincon/EDTA
Type Copper/Zinc Tablet
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.