missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Lovibond™ Total Nitrogen Reagent Set, for Tube Testing, Low Range
Description
Includes
Total N Persulfate Reagent, Total Nitrogen Reagent A, Total Nitrogen Reagent B, Total Nitrogen Hydroxide LR Tubes, Total Nitrogen Acid LR/HR Tubes, Deionized Water, 100mL
- Offers same high quality standards as tablet reagents
- Chemical properties are suitable for use with Hach™ colorimeters, spectrophotometers and test kits
- Simply tear open Powder Pack and pour reagent into sample
- Large design makes them easier to handle
- “Easy Open” notch makes it simple to tear open package
- Easily create “V”, to allow user to pour reagent into sample
- Beveled edge prevents reagent from getting stuck in corners
Municipal Drinking Water and Wastewater Testing; Industrial Wastewater and Process Control Testing; Environmental Water Testing
Specifications
Specifications
| For Use With (Application) | Municipal drinking water and wastewater testing, industrial wastewater and process control testing, environmental water testing |
| For Use With (Equipment) | Hach™ and other manufacturers test kits, colorimeters and spectrophotometers, reactor |
| Includes | Total N persulfate reagent (50 pack), total nitrogen reagent A (50 pack), total nitrogen reagent B (50 pack), 50 ct. total nitrogen hydroxide LR tubes, 50 ct. total nitrogen acid LR/HR tubes, 100mL deionized water |
| No. of Tests | 25 |
| Range | 0 to 25mg/L |
| Test Method | Persulfate Digestion |
| Type | Low Range Total Nitrogen Reagent Set |
Safety and Handling
Product Identifier
- 50 UNIT VARIO TOTAL NITROGEN LR REAGENT SET .
- Danger
- Acute toxicity Category 4
- Serious eye damage/eye irritation Category 1
- Serious eye damage/eye irritation Category 2
- Substances and mixtures corrosive to metals Category 1
- Oxidising solid Category 3
- Respiratory sensitiser Category 1
- Skin corrosion/irritation Category 1 A
- Skin corrosion/irritation Category 2
- Skin sensitiser Category 1
- Specific target organ toxicity Category 3
- H272-May intensify fire; oxidiser.
- H290-May be corrosive to metals.
- H302-Harmful if swallowed.
- H314-Causes severe skin burns and eye damage.
- H315-Causes skin irritation.
- H317-May cause an allergic skin reaction.
- H318-Causes serious eye damage.
- H319-Causes serious eye irritation.
- H334-May cause allergy or asthma symptoms or breathing difficulties if inhaled.
- H335-May cause respiratory irritation.
- EUH031-Contact with acids liberates toxic gas.
- P260-Do not breathe dust/fume/gas/mist/vapours/spray.
- P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P301+P310-IF SWALLOWED: Immediately call a POISON CENTER/doctor /
- P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.
- P302+P352-IF ON SKIN: Wash with plenty of water/.
- P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P308+P313-IF exposed or concerned: Get medical advice/attention.
- P310-Immediately call a POISON CENTER/doctor/.
- P342+P311-If experiencing respiratory symptoms: Call a POISON CENTER/doctor/ .
- P390-Absorb spillage to prevent material damage.
- P501b-Dispose of contents/container in accordance with local/regional/national/international regulations.
- MIXTURE LIST-Contains : disodium peroxodisulphate, potassium persulphate, disodium disulphite, disodium4,5-dihydroxynapthal
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?