missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Lovibond™ Silica Reagent Set, Powder Packs™, High Range
Click to view available options
No. of Tests:
100
Unit Size:
10mL
Description
Includes
Silica HR Acid Reagent (10mL sample, 100 pack), Silica HR Citric Acid Reagent (10mL sample, 100 pack), Silica HR Molybdate Reagent (10mL sample, 100 pack)
- Offers same high quality standards as tablet reagents
- Chemical properties are suitable for use with Hach™ colorimeters, spectrophotometers and test kits
- Simply tear open Powder Pack and pour reagent into sample
- Large design makes them easier to handle
- “Easy Open” notch makes it simple to tear open package
- Easily create “V”, to allow user to pour reagent into sample
- Beveled edge prevents reagent from getting stuck in corners
Municipal Drinking Water and Wastewater Testing; Industrial Wastewater and Process Control Testing; Environmental Water Testing
Specifications
Specifications
| For Use With (Application) | Municipal drinking water and wastewater testing, industrial wastewater and process control testing, environmental water testing |
| For Use With (Equipment) | Hach™ and other manufacturers test kits, colorimeters and spectrophotometers |
| Includes | 10mL silica HR acid reagent (100 pack), 10mL silica HR citric acid reagent (100 pack), 10mL silica HR molybdate reagent (100 pack) |
| No. of Tests | 100 |
| Range | 0 to 100mg/L |
| Test Method | Salicylate Silicomolybdate |
| Type | High Range Silica Reagent Set |
Safety and Handling
Product Identifier
- vario silica HR set
- Warning
- Serious eye damage/eye irritation Category 2
- Skin corrosion/irritation Category 2
- LONG-TERM AQUATIC HAZARD Chronic 3
- H319-Causes serious eye irritation.
- H315-Causes skin irritation.
- H412-Harmful to aquatic life with long lasting effects.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P273-Avoid release to the environment.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P311-Call a POISON CENTER/doctor/ .
- P302+P352-IF ON SKIN: Wash with plenty of water/.
- P332+P313-If skin irritation occurs: Get medical advice/attention.
- P337+P313-If eye irritation persists: Get medical advice/attention.
- MIXTURE LIST-Contains : sulphamic acid, citric acid
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction