Learn More
Lovibond™ Primary Chlorine Standard Kit for Colorimeters, Cole-Parmer™
Shop All Cole-Parmer ProductsDescription
- Complies with new regulations specified in US EPA Method 334.0.
- Ideal for laboratory and on-line chlorine measurements.
- Chlorine standard ampoule, 97mL of zero chlorine water and ampoule breaker.
- Everything you need for easy preparation of a 1.5mg/L NIST traceable chlorine standard.
- Simple, time saving and practical kit to help comply with the new regulations
Specifications
Specifications
| Description | Chlorine standard ampoule, 97mL of zero chlorine water, and ampoule breaker |
| Packaging | 1Box/Kit |
| For Use With (Equipment) | Colorimeter |
| Quantity | 1 Box/Kit |