missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Lovibond™ Nitrite 3 Powder Packs™: for Tube Testing, Low Range
Click to view available options
Range:
0 to 0.3mg/L
Unit Size:
Each
Description
- Offers same high quality standards as tablet reagents
- Chemical properties are suitable for use with Hach™ colorimeters, spectrophotometers and test kits
- Simply tear open Powder Pack and pour reagent into sample
- Large design makes them easier to handle
- “Easy Open” notch makes it simple to tear open package
- Easily create “V”, to allow user to pour reagent into sample
- Beveled edge prevents reagent from getting stuck in corners
Municipal Drinking Water and Wastewater Testing; Industrial Wastewater and Process Control Testing; Environmental Water Testing
Specifications
Specifications
| Certifications/Compliance | US EPA |
| Color | Light Beige |
| Composition | Potassium Dihydrogenorthophosphate 75 to 85%, Sodium Sulfanilite Dihydrate 5 to 10%, Potassium Disulfate 1 to 10%, non-hazardous components 1 to 5% |
| pH Range | 3.1 |
| Volume (Metric) | 10 mL |
| Format | Powder |
| For Use With (Application) | Municipal drinking water and wastewater testing, industrial wastewater and process control testing, environmental water testing |
| For Use With (Equipment) | Hach™ and other manufacturers test kits, colorimeters and spectrophotometers |
| Range | 0 to 0.3mg/L |
| Type | Nitrite 3 Reagent |
| Show More |
Safety and Handling
Product Identifier
- Nitrite LR VARIO Powder Pack .
- Danger
- Serious eye damage/eye irritation Category 1
- Skin corrosion/irritation Category 1 A
- H314-Causes severe skin burns and eye damage.
- H318-Causes serious eye damage.
- P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P310-Immediately call a POISON CENTER/doctor/.
- MIXTURE LIST-Contains : disodium 4,5-dihydroxynapthalene-2,7-disulphonate
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction