missing translation for 'onlineSavingsMsg'
Learn More

Lovibond™ Powder Pack™ Reagents: Iron Ferro; 10mL

Product Code. 10691694
Click to view available options
Range:
0.02 to 3mg/L
Unit Size:
Pack of 100
This item is not returnable. View return policy

Product Code. 10691694

Brand: Lovibond™ 530560

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Pre-measured, unit dose reagent for water quaility testing

  • Simply tear open Powder Pack and pour reagent into sample
  • Large design makes them easier to handle
  • “Easy Open” notch makes it simple to tear open the package
  • Easily create a “V”, to allow user to pour reagent into sample
  • Beveled edge prevents reagent from getting stuck in the corners

Municipal Drinking Water and Wastewater Testing; Industrial Wastewater and Process Control Testing; Environmental Water Testing

Compliance

Iron-Ferro reagent is US EPA accepted

Specifications

Certifications/Compliance US EPA
Composition Sodium Dithionite 30 to 40%, Disodium Disulphite 20 to 30%, Sodium Thiosulphate 30 to 40%, 1, 10-phenanthroline Monohydrochloride Monohydrate 1 to 5%, Sodium Citrate tribasic Dihydrate 1 to 10%
Volume (Metric) 10 mL
For Use With (Application) Municipal drinking water and wastewater testing, industrial wastewater and process control testing, environmental water testing
For Use With (Equipment) Test kits, colorimeters and spectrophotometers
Range 0.02 to 3mg/L
Test Method 1,10-Phenanthroline
Type Iron Ferro Reagent
Volume (English) 0.33 oz.
compliance-icons compliance-icons
Product Identifier
  • Vario Ferro F10, 25Ml
Signal Word
  • Danger
Hazard Category
  • Serious eye damage/eye irritation Category 1
  • Acute toxicity Category 4
  • LONG-TERM AQUATIC HAZARD Chronic 3
Hazard Statement
  • H302-Harmful if swallowed.
  • H318-Causes serious eye damage.
  • H412-Harmful to aquatic life with long lasting effects.
  • EUH031-Contact with acids liberates toxic gas.
  • EUH208-Contains . May produce an allergic reaction.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P310-IF SWALLOWED: Immediately call a POISON CENTER/doctor /
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Supplemental information
  • MIXTURE LIST-Contains : Sodium dithionite, disodium disulphite,1, 10-phenanthroline, methenamine
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.