missing translation for 'onlineSavingsMsg'
Learn More

Lovibond™ MINIKITS Test Kits: Quaternary Ammonium Compounds

Product Code. 10686911
Change view
Click to view available options
No. of Tests:
50
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. No. of Tests Unit Size
10686911 50 Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 10686911 Supplier Lovibond™ Supplier No. 414170
  • ONLINE EXCLUSIVE PRICE
    £69.30
    £65.90 / Pack of 1
    Save £3.40
    4% Off
View all promotions
 Restricted Item
Estimated Shipment: 29-12-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Includes

  • Calibrated Sample Container, 100mL
  • QAC-Test Tablets (200 tablets)
  • Instructions
  • Storage Case

Accurate test kits for routine water analysis

  • Uses titrimetric method for determination of QAC
  • Easy operation and exact reagent dosing
  • Foil-wrapped tablet reagents with minimum guaranteed shelf life of five years
  • Easy and safe storage

Specifications

For Use With (Application) Routine water analysis
Includes 100mL calibrated sample container, QAC-test tablets, instructions, storage case
No. of Tests 50
Range 0 to 80mg/L
Type Quaternary Ammonium Compounds Test Kit
compliance-icons compliance-icons
Product Identifier
  • MINIKIT TEST QAC TEST AF417
Signal Word
  • Danger
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Reproductive toxicity Category 1B
Hazard Statement
  • H319-Causes serious eye irritation.
  • H360-May damage fertility or the unborn child.
Precautionary Statement
  • P201-Obtain special instructions before use.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P308+P313-IF exposed or concerned: Get medical advice/attention.
  • P405-Store locked up.
Supplemental information
  • MIXTURE LIST-Contain: boric acid, sodium dodecyl sulphate
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.