Learn More
Lovibond™ DPD No. 1 HR Tablets
Determine the amount of free chlorine in your hot tub, swimming pool, or spa water with these foil-packaged, fast-acting tablets. Lovibond™ DPD No. 1 HR Tablets are foil-packaged tablets for consistent and reliable reagent delivery.
Brand:  Lovibond™ 511500BT
Product Code. 15123058
  • £21.00 / 100 tablets
Estimated Shipment: 31-10-2024
to see stock.
Explore more special offers
Add to basket
Description
Description
Determine the amount of free chlorine in your hot tub, swimming pool, or spa water with these foil-packaged, fast-acting tablets. Lovibond™ DPD No. 1 HR Tablets are foil-packaged tablets for consistent and reliable reagent delivery.
  • Simple, time-saving and practical
  • Foil packaged test tablets remain the most consistent and reliable reagent delivery system available
  • Each tablet is hermetically sealed within an individually formed foil cup
Specifications
Specifications
White
6.3
Free chlorine analysis in water
0.01 to 10ppm
DPD
N,N-diethylbenzene-1,4-diammonium Sulphate 2.5 to 5%, Citric Acid 0.1 to 1%, non-hazardous components 20 to 35%, Sodium cyclamate 60 to 70%, Polyvinyl Alcohol ≤0.1%, Di-Sodium Dihydrogenethylenediaminetetraacetate ≤2.5%
Tablet
SC450 chlorine 2 and 3 colorimeters, MC500 colorimeter, SP600 spectrophotometer, AquaPRO3 and PRO6 Pool photometers
20°C
DPD No. 1 HR Tablet