missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Lovibond™ DPD No. 1 High Calcium Tablets
Specifications
Specifications
| Color | White |
| Composition | N,N-diethylbenzene-1,4-diammonium Sulphate 0.1 to 1%, non-hazardous components >99% |
| pH Range | 6.2 |
| Format | Tablet |
| For Use With (Application) | Water analysis |
| For Use With (Equipment) | SC450 chlorine 2 and 3 colorimeters, MC500 colorimeter, SP600 spectrophotometer |
| Range | 0.05 to 13mg/L |
| Storage Requirements | 20°C |
| Test Method | DPD |
| Type | DPD No. 1 High Calcium Tablet |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?