Learn More
Lovibond™ Colorimeter Reagent Set: Ozone
For use with MC500, MD600 colorimeter and SP600 spectrophotometer.
Brand: Lovibond™ 517791BT
Code : NEW
Additional Details : Weight : 0.30000kg
Includes: 100 each of DPD No. 1 tablets, DPD No. 3 tablets and glycine tablets
Description
For use with MC500, MD600 colorimeter and SP600 spectrophotometer.Specifications
Ozone Colorimeter Reagent Set | |
MC500, MD600 colorimeter and SP600 spectrophotometer | |
100 each of DPD No. 1 tablets, DPD No. 3 tablets and glycine tablets |