Learn More
Lovibond™ Colorimeter: Molybdate
These rugged, waterproof, single parameter colorimeters are ideal for laboratory and field testing
Brand: Lovibond Tintometer 276140
Code : NEW
Additional Details : Weight : 0.78500kg
Description
- Range: 0.03 - 3.0mg/L
- Method: Molybdate
- Reagent set RS5450 included (100 tests)
- Large, backlit display; real time clock; data logging capabilities
- Ability to export data using the IRiM
Specifications
3% Full scale | |
CE | |
Backlit LCD | |
IP68 | |
Molybdate Colorimeter | |
1.5 in. | |
Three 24mm sample cells with light-shield caps, reagents for 100 tests, Foam-lined case and Four AAA batteries | |
15.5 cm | |
0.01 A | |
2 Years | |
250 g | |
7.6 cm |
Yes | |
Automatic; 16 data sets with time and date stamp | |
6.1 x 3 x 1.5 in. (15.5 x 7.6 x 3.8 cm) | |
LED | |
Laboratory and field testing | |
3.8 cm | |
6.1 in. | |
0.03 to 3 mg/L | |
5°C to 40°C | |
0.55 lb. | |
3 in. |
Safety and Handling
- PHOTOMETRE MD100 molybdene PP LR 0,03-3 MG/L
Signal Word
- Danger
Hazard Category
- Acute toxicity Category 4
- LONG-TERM AQUATIC HAZARD Chronic 3
- ACUTE AQUATIC Acute 1
- Serious eye damage/eye irritation Category 1
- Serious eye damage/eye irritation Category 2
- Respiratory sensitiser Category 1
- Skin corrosion/irritation Category 2
- Skin sensitiser Category 1
- Specific target organ toxicity Category 3
Hazard Statement
- H302-Harmful if swallowed.
- H315-Causes skin irritation.
- H317-May cause an allergic skin reaction.
- H318-Causes serious eye damage.
- H319-Causes serious eye irritation.
- H334-May cause allergy or asthma symptoms or breathing difficulties if inhaled.
- H335-May cause respiratory irritation.
- H400-Very toxic to aquatic life.
- H412-Harmful to aquatic life with long lasting effects.
Precautionary Statement
- P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
- P264-Wash thoroughly after handling.
- P273-Avoid release to the environment.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P302+P352-IF ON SKIN: Wash with plenty of water/.
- P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P308+P313-IF exposed or concerned: Get medical advice/attention.
- P310-Immediately call a POISON CENTER/doctor/.
- P333+P313-If skin irritation or rash occurs: Get medical advice/attention.
- P337+P313-If eye irritation persists: Get medical advice/attention.
- P501b-Dispose of contents/container in accordance with local/regional/national/international regulations.
Supplemental information
- MIXTURE LIST-Contains : octylphenol polyethoxyethanol, cetrimonium bromiide, succinic acid, glycine, disodium peroxodisulpha