Learn More
Lovibond™ Color of Varnish (Gardner Color) Discs
Brand:  Lovibond™ 243040
Code : NEW
Additional Details : Weight : 0.25000kg
 View more versions of this product
Product Code. 15843032
  • £355.00 / Each
Estimated Shipment: 04-07-2024
to see stock.
Specifications
ASTM D-1544, AOCS Td 1a
Color of Varnish l Disc
Varnish comparator kits 705-V2, 705-VA2 or 705-VA4
9 to 18