missing translation for 'onlineSavingsMsg'
Learn More

Lovibond™ Chloride T2 Tablets

Product Code. 15175894
Click to view available options
Range:
0.5 to 25mg/L
Unit Size:
Pack of 100
This item is not returnable. View return policy

Product Code. 15175894

Brand: Lovibond™ 515920BT

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Lovibond Chloride T2 Tablets, Range: 0.5 to 25 mg/L, White, Composition: Potassium Monopersulfate Triple salt 5 to 10%, Silver Nitrate <2.5%, Boric Acid >50%, non-hazardous components <20%, Format: Tablet, For Use With: Water analysis, MD600 colorimeter and the spectrodirect spectrophotometer

Lovibond Chloride T2 Tablets, Range: 0.5 to 25mg/L, White, Composition: Potassium Monopersulfate Triple salt 5 to 10%, Silver Nitrate <2.5%, Boric Acid >50%, non-hazardous components <20%, Format: Tablet, For Use With: Water analysis, MC500 colorimeter and SP600 spectrophotometer

Specifications

Color White
Composition Potassium Monopersulfate Triple salt 5 to 10%, Silver Nitrate <2.5%, Boric Acid >50%, non-hazardous components <20%
pH Range 4
Format Tablet
For Use With (Application) Water analysis
For Use With (Equipment) MC500 colorimeter and SP600 spectrophotometer
Range 0.5 to 25mg/L
Storage Requirements 20°C
Test Method Silver Nitrate/Turbidity
Type Chloride T2 Tablet
compliance-icons compliance-icons
Product Identifier
  • Chloride T2
Signal Word
  • Danger
Hazard Category
  • Skin corrosion/irritation Category 1 B
  • Serious eye damage/eye irritation Category 1
  • ACUTE AQUATIC Acute 1
  • LONG-TERM AQUATIC HAZARD Chronic 1
Hazard Statement
  • H314-Causes severe skin burns and eye damage.
  • H410-Very toxic to aquatic life with long lasting effects.
Precautionary Statement
  • P273-Avoid release to the environment.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.
  • P501b-Dispose of contents/container in accordance with local/regional/national/international regulations.
Supplemental information
  • MIXTURE LIST-Contains : potassium monopersulfate triple salt, silver nitrate
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.