Learn More
Lovibond™ CHECKIT™ Comparator Test Kits: Total Hardness LR
Accurate test kits for routine water analysis
Brand: Lovibond™ 414260
Code : 04
Additional Details : Weight : 0.26600kg Transport : UN number : 3259 Chem class : 8 Pack group : III
Includes:
- Calibrated sample container, 100mL
- Total Hardness VLR Tablets (250 tablets)
- Instructions
- Storage Case
Specifications
Routine water analysis | |
100 | |
VLR Total Hardness Mini Kit |
100mL calibrated sample container, total hardness VLR tablets, instructions, storage case | |
1 to 10mg/L |
Safety and Handling
- MINIKIT TEST durete VERY LOW AF426
Signal Word
- Warning
Hazard Category
- Serious eye damage/eye irritation Category 2
- Skin corrosion/irritation Category 2
Hazard Statement
- H315-Causes skin irritation.
- H319-Causes serious eye irritation.
Precautionary Statement
- P264-Wash thoroughly after handling.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P302+P352-IF ON SKIN: Wash with plenty of water/.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Supplemental information
- MIXTURE LIST-Contains : 2-amino-2-methylpropane-1,3-diol, tri-sodium phosphate dodecahydrate