missing translation for 'onlineSavingsMsg'
Learn More

Lovibond™ CHECKIT™ Comparator Test Kits: Total Hardness HR

Product Code. 10300071
Change view
Click to view available options
Range:
20 to 800mg/L
5 to 500mg/L
Unit Size:
Each
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Range unitSize
10300071 5 to 500mg/L Each
10064500 20 to 800mg/L Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10300071 Supplier Lovibond™ Supplier No. 414240

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Accurate test kit for routine water analysis

Specifications

For Use With (Application) Routine water analysis
Includes 30mL calibrated sample container, 30mL calibrated sample container, total hardness tablets, instructions, storage case
No. of Tests 100
Range 5 to 500mg/L
Type Total Hardness Mini Kit
compliance-icons compliance-icons
Product Identifier
  • total hardness
Signal Word
  • Danger
Hazard Category
  • Serious eye damage/eye irritation Category 1
  • Skin corrosion/irritation Category 2
Hazard Statement
  • H315-Causes skin irritation.
  • H318-Causes serious eye damage.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.
Supplemental information
  • MIXTURE LIST-Contain: lithium hydroxide
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.