missing translation for 'onlineSavingsMsg'
Learn More

Lovibond™ CHECKIT™ Comparator Test Kits: Aluminum

Accurate test kits for routine water analysis

Brand:  Lovibond™ 147200


Product Code. 10204792

  • £69.00 / Each

Please to purchase this item. Need a web account? Register with us today!

Explore more special offers

Includes:
  • Aqua Comparator Housing
  • Aqua Comparator Disc
  • Aluminum No. 1 Tablets (100 pack)
  • Aluminum No. 2 Tablets (100 pack)
  • 2 x 13.5mm Sample Cells
  • Tablet Crusher
  • Instructions
  • Carrying Case
This item is not returnable. View return policy

Description

Description

  • Easy to use: add reagent to sample cell, rotate disc until color matches sample, and read concentration value

Specifications

Specifications

±5%
Aqua comparator™ housing, aqua comparator™ disc, aluminum No. 1 tablets, aluminum No. 2 tablets, 2 x 13.5mm sample cells, tablet crusher, instructions, carrying case
0 to 0.3mg/L
Routine water analysis
100
Aluminum Test Kit
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

Product Identifier
  • comparitor aluminium 0-0.3mg/l

Signal Word
  • Warning

Hazard Category
  • Flammable solid Category 2
  • Skin sensitiser Category 1

Hazard Statement
  • H228-Flammable solid.
  • H317-May cause an allergic skin reaction.

Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P333+P313-If skin irritation or rash occurs: Get medical advice/attention.

Supplemental information
  • MIXTURE LIST-Contains : methenamine

SDS
Documents

Documents

Certificates
Special Offers

Special Offers