missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Lovibond™ CHECKIT™ Comparator Test Kits
Description
Includes
Carrying case, comparator, color disc, sample cells, instructions and enough reagents for 100 tests
- Easy to use: add reagent to sample cell, rotate disc until color matches sample, and read concentration value
- Color discs and reagents are available separately
Specifications
Specifications
| For Use With (Application) | For routine water analysis |
| Range | 0-5.0mg/L |
| Sufficient For | 100 Tests |
| Type | Copper |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?