missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Lovibond™ Borosilicate Glass Colorimetry Cells
Precision fused cells
£63.06 - £68.25
Product Code | Brand | For Use With (Equipment) | Pathlength | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | For Use With (Equipment) | Pathlength | Price | Quantity & Availability | ||||
16536640
|
Lovibond™
655960 |
Model Fs, PFX195, 880, 950 and 995 instruments with heated Samples (up to 100°C) | 10 mm |
£63.06
Each |
Estimated Shipment: 14-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | |||
15879791
|
Lovibond™
656080 |
Model Fs, PFX195, 880, 950 and 995 instruments with heated Samples (up to 100°C) | 2.54 cm |
£68.25
Each |
Estimated Shipment: 04-07-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | |||
Description
Available in a range of dimensions and path lengths, made from optical glass to the highest standards. Note: For instruments equipped with a heater unit and whenever cells are subjected to thermal shock, it is recommended that borosilicate cells be used.
Series: W600 Borosilicate
Specifications
Colorimetry Cell | |
Borosilicate Glass |