missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Lovibond™ Alka-M High Range Colorimtere Tablets Reagent
Click to view available options
Range:
5 to 500ppm
Unit Size:
Pack of 100
Description
Lovibond Alka-M High Range Photometer Tablets, Alka-M HR Tablet, Range: 5 to 500ppm, Orange, Composition: Adipic Acid 10 to 20%, non-hazardous components 80 to 90%, Format: Tablet, For Use With: Water analysis, MC500 colorimeter, SP600 spectrophotometer, AquaPRO6, Method: Acid Indicator
Specifications
Specifications
| Color | Orange |
| Composition | Adipic Acid 10 to 20%, non-hazardous components 80 to 90% |
| pH Range | 3.1 |
| Format | Tablet |
| For Use With (Application) | Water analysis |
| For Use With (Equipment) | MC500 colorimeter, SP600 spectrophotometer, AquaPRO6 |
| Range | 5 to 500ppm |
| Storage Requirements | 20°C |
| Test Method | Acid Indicator |
| Type | Alka-M HR Tablet |
Safety and Handling
Product Identifier
- X100 PASTILLE ALCALINITE - M HR
- Warning
- Serious eye damage/eye irritation Category 2
- H319-Causes serious eye irritation.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P337+P313-If eye irritation persists: Get medical advice/attention.
- MIXTURE LIST-Contains : adipic acid
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction