missing translation for 'onlineSavingsMsg'
Learn More

Lovastatin, MedChemExpress

Product Code. 16449096
Encompass_Preferred
Change view
Click to view available options
Quantity:
10 mg
50 mg
100 mg
Unit Size:
100mg
10mg
50mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16449096 100 mg 100mg
16434847 50 mg 50mg
16488998 10 mg 10mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 16449096 Supplier Medchem Express Supplier No. HYN0504
 Restricted Item
Estimated Shipment: 19-10-2026
Add to basket
Add to basket
This item is not returnable. View return policy

MedChemExpress Lovastatin is a cell-permeable HMG-CoA reductase inhibitor used to lower cholesterol.

Lovastatin is a cell-permeable HMG-CoA reductase inhibitor used to lower cholesterol.

Specifications

Chemical Name or Material Lovastatin
CAS 75330-75-5
Color White
Purity Grade Notes Research
Molecular Formula C24H36O5
Quantity 100 mg
Synonym Mevinolin
Solubility Information DMSO : ≥ 100 mg/mL (247.19 mM)
SMILES O=C([C@@H](C)CC)O[C@@H]1[C@@]([C@H]2CC[C@@H](OC3=O)C[C@H](C3)O)([H])C(C=C[C@@H]2C)=C[C@H](C)C1
Molecular Weight (g/mol) 404.54
Formula Weight 404.54
Percent Purity 99.89%
Grade Research
Physical Form Solid
For Use With (Application) COVID-19-immunoregulation
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.