missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals Lovastatin, 98%

Product Code. 15500594 Shop All Thermo Scientific Chemicals Products
Click to view available options
Quantity:
5 g
25 g
100 g
Unit Size:
100g
25g
5g
This item is not returnable. View return policy

Product Code. 15500594

Brand: Thermo Scientific Chemicals 458830250

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Lovastatin, CAS # 75330-75-5, is a cholesterol lowering compound and inhibitor of HMG-CoA reductase, a rate limiting enzyme in cholesterol synthesis. | CAS: 75330-75-5 | C24H36O5 | 404.55 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

General Description

Applications

• It blocks product ion of mevalonate, a critical compound in the production of cholesterol and isoprenoids

• It can increase cellular resistance to anticancer agents such as doxorubicin • It can induce apoptosis in myeloma cells

• It can cause cell cycle arrest in G1 and G2/M phase

• It also blocks PDGF receptor tyrosine phosphorylation and MAP kinase activation by PDGF

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 75330-75-5
Molecular Formula C24H36O5
Molecular Weight (g/mol) 404.55
MDL Number MFCD00072164
InChI Key PCZOHLXUXFIOCF-BXMDZJJMSA-N
Synonym lovastatin, mevinolin, mevacor, monacolin k, lovalip, altoprev, lovalord, mevinacor, nergadan, 6alpha-methylcompactin
PubChem CID 53232
ChEBI CHEBI:40303
IUPAC Name [(1S,3R,7S,8S,8aR)-8-[2-[(2R,4R)-4-hydroxy-6-oxooxan-2-yl]ethyl]-3,7-dimethyl-1,2,3,7,8,8a-hexahydronaphthalen-1-yl] (2S)-2-methylbutanoate
SMILES CC[C@H](C)C(=O)O[C@H]1C[C@@H](C)C=C2C=C[C@H](C)[C@H](CC[C@@H]3C[C@@H](O)CC(=O)O3)[C@@H]12

Specifications

Melting Point 175.0°C
Infrared Spectrum Authentic
Assay Percent Range 97.5% min. (HPLC)
Quantity 25 g
Solubility Information Solubility in water: insoluble. Other solubilities: less soluble in ethanol, soluble in chloroform and acetone
Formula Weight 404.54
Percent Purity 98%
Chemical Name or Material Lovastatin
compliance-icons compliance-icons
Product Identifier
  • Lovastatin
Signal Word
  • Warning
Hazard Category
  • Carcinogenicity Category 2
  • Reproductive toxicity Category 2
  • Respiratory sensitiser Category 1
Hazard Statement
  • H351-Suspected of causing cancer.
  • H361d-Suspected of damaging the unborn child.
  • H317-May cause an allergic skin reaction.
Precautionary Statement
  • P201-Obtain special instructions before use.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P308+P313-IF exposed or concerned: Get medical advice/attention.

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.