missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Lotus Tetragonolobus (Asparagus Pea) Lectin (LTL), fluorescein (FITC)

Product Code. 17146642
Change view
Click to view available options
Quantity:
2 mg
Unit Size:
2mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
17146642 2 mg 2mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 17146642 Supplier Invitrogen™ Supplier No. L32480
 Restricted Item
Estimated Shipment: 13-11-2026
Add to basket
Edge
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Bright-green LTL-fluorophor conjugate

  • Lotus Tetragonolobus Lectin (LTL), fluorescein (FITC), is a bright-green LTL-fluorophor conjugate. LTL is isolated from Tetragonolobus purpureus, also known as Lotus tetragonolobus, within the common names asparagus pea or winged pea.
  • Combined with other lectins, it has been used for the detection of certain diseased tissue in kidney and the detection of colon cancer. LTL has also been used in studies of the inhibition of macrophage migration and in comparison to macrophage inhibition factor (MIF).
  • Very bright FITC conjugate detected with traditional FITC/GFP filter set, Ex/Em: 495/515 nm
  • Applications: immunofluorescence, glycobiology
  • Sugar specificity: fucose, arabinose
  • Inhibiting/eluting sugar: 50–100 mM L-fucose
  • Blood group specificity: non-specific, O<A2
  • Unconjugated lectin molecular weight: 107 KDa
  • LTL-FITC is provided at a 2 mg/mL concentration in 10 mM HEPES, 0.15 M NaCl, pH 7.5, 0.08% sodium azide. 0.1 mM Ca2+
  • A precipitate may form during storage. This does not have a significant adverse effect on the product. If a precipitate forms upon long-term storage at 4°C, warm to 37°C and centrifuge before use.
TRUSTED_SUSTAINABILITY

Specifications

Quantity 2 mg
Source Asparagus pea
Conjugate FITC
Content And Storage When stored at or below 2°C to 8°C in the dark, the product is stable for 6 months after reciept.
Label Type Classic Dyes
Shipping Condition Approved for shipment at Room Temperature or on Wet Ice
Form Solution
Protein Subtype Lectins

For Research Use Only. Not for use in diagnostic procedures.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.