missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals Losartan potassium salt

Product Code. p-7139371 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
5 g
25 g
Unit Size:
25g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15831052 5 g 5g
15841052 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 15831052 Supplier Thermo Scientific Chemicals Supplier No. 462740050
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 124750-99-8 | C22H22ClKN6O | 461.01 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 124750-99-8
Molecular Formula C22H22ClKN6O
Molecular Weight (g/mol) 461.01
InChI Key OXCMYAYHXIHQOA-UHFFFAOYSA-N
PubChem CID 53396294

Specifications

Quantity 5 g
Chemical Name or Material Losartan potassium salt

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.