missing translation for 'onlineSavingsMsg'
Learn More

Losartan Potassium Salt, TRC

Product Code. 30132792
Change view
Click to view available options
Quantity:
10 mg
5 g
25 g
Unit Size:
10mg
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30132792 25 g 25g
30009193 5 g 5g
18012210 10 mg 10mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30132792 Supplier Toronto Research Chemicals Supplier No. TRCL47050025G
 Restricted Item
Estimated Shipment: 08-10-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 124750-99-8
Molecular Formula C22 H22 Cl N6 O . K
Molecular Weight (g/mol) 461
Synonym Losartan potassium, Potassium 5-[4'-[[2-butyl-4-chloro-5-(hydroxymethyl)-1H-imidazol-1-yl]methyl]biphenyl-2-yl]tetrazol-1-ide
IUPAC Name potassium;[2-butyl-5-chloro-3-[[4-[2-(1,2,3-triaza-4-azanidacyclopenta-2,5-dien-5-yl)phenyl]phenyl]methyl]imidazol-4-yl]methanol
SMILES [K+].CCCCc1nc(Cl)c(CO)n1Cc2ccc(cc2)c3ccccc3c4nnn[n-]4

Specifications

Quantity 25 g
InChI Formula InChI=1S/C22H22ClN6O.K/c1-2-3-8-20-24-21(23)19(14-30)29(20)13-15-9-11-16(12-10-15)17-6-4-5-7-18(17)22-25-27-28-26-22;/h4-7,9-12,30H,2-3,8,13-14H2,1H3;/q-1;+1
Formula Weight 460.1181
Percent Purity >95%
Chemical Name or Material Losartan Potassium
Grade HPLC
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.