missing translation for 'onlineSavingsMsg'
Learn More

Losartan potassium, MedChemExpress

Product Code. 16471689
Encompass_Preferred
Change view
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16471689 1 g 1g
16499847 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 16471689 Supplier Medchem Express Supplier No. HY17512A
 Restricted Item
Estimated Shipment: 25-09-2026
Add to basket
Add to basket
This item is not returnable. View return policy

MedChemExpress Losartan potassium (DuP-753 potassium) is an angiotensin II receptor type 1 (AT1) antagonist, competing with the binding of angiotensin II to AT1 with an IC50 of 20 nM.

Losartan potassium (DuP-753 potassium) is an angiotensin II receptor type 1 (AT1) antagonist, competing with the binding of angiotensin II to AT1 with an IC50 of 20 nM.

Specifications

Chemical Name or Material Losartan potassium
CAS 124750-99-8
Color White
Purity Grade Notes Research
Molecular Formula C22H22ClKN6O
Quantity 1 g
Synonym DuP-753 potassium
Solubility Information DMSO : ≥ 110 mg/mL (238.61 mM) ∣H2O : 50 mg/mL (108.46 mM; Need ultrasonic)
SMILES OCC1=C(Cl)N=C(CCCC)N1CC2=CC=C(C3=CC=CC=C3C4=N[N-]N=N4)C=C2.[K+]
Molecular Weight (g/mol) 461.0
Formula Weight 461.0
Percent Purity 99.57%
Grade Research
Physical Form Solid
For Use With (Application) COVID-19-immunoregulation
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.