missing translation for 'onlineSavingsMsg'
Learn More

Losartan EP Impurity J (Losartan USP Related Compound B), Mikromol™

Product Code. 30463793
Change view
Click to view available options
Quantity:
25 mg
100 mg
4 x 25 mg
Unit Size:
100mg
25mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30463793 4 x 25 mg -
30049658 100 mg 100mg
30051065 25 mg 25mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30463793 Supplier Mikromol Supplier No. MM0168.1800254AMP
 Restricted Item
Estimated Shipment: 03-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy

Discover Mikromol - your trusted source for high-quality pharmaceutical reference standards. Supporting accurate, compliant analysis with ISO 17034-accredited materials designed for confidence in every result.

Losartan EP Impurity J (Losartan USP Related Compound B), Mikromol
TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material O-Acetyl Losartan
CAS 1006062-27-6
Analyte or Component Names O-Acetyl Losartan
Shipping Condition Room Temperature
Molecular Formula C24 H25 Cl N6 O2
Certifications/Compliance ISO 17025
Synonym [2-Butyl-4-chloro-1-[[2'-(1H-tetrazol-5-yl)biphenyl-4-yl]methyl]-1H-imidazol-5-yl]methyl Acetate, Losartan Potassium Imp. J (EP), Losartan Imp. J (EP), Losartan USP Related Compound B, Losartan USP RC B, (2-Butyl-4-chloro-1-[[2'-(1H-tetrazol-5-yl)biphenyl-4-yl]methyl]-1H-imidazol-5-yl)methyl Acetate, Losartan Related Compound B, Losartan Potassium Impurity J, Losartan Impurity J
InChI Formula InChI=1S/C24H25ClN6O2/c1-3-4-9-22-26-23(25)21(15-33-16(2)32)31(22)14-17-10-12-18(13-11-17)19-7-5-6-8-20(19)24-27-29-30-28-24/h5-8,10-13H,3-4,9,14-15H2,1-2H3,(H,27,28,29,30)
Solution Type Neat
SMILES CCCCc1nc(Cl)c(COC(=O)C)n1Cc2ccc(cc2)c3ccccc3c4nnn[nH]4
IUPAC Name [2-butyl-5-chloro-3-[[4-[2-(1H-tetrazol-5-yl)phenyl]phenyl]methyl]imidazol-4-yl]methyl acetate
Molecular Weight (g/mol) 464.95
Quantity 4 x 25 mg
Formula Weight 464.1728 g/mol
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.