missing translation for 'onlineSavingsMsg'
Learn More

Lornoxicam, TRC

Product Code. 18093410
Change view
Click to view available options
Quantity:
10 mg
50 mg
250 mg
Unit Size:
10mg
250mg
50mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
18093410 10 mg 10mg
18000620 50 mg 50mg
18032620 250 mg 250mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 18093410 Supplier Toronto Research Chemicals Supplier No. TRCL47000010MG
 Restricted Item
Estimated Shipment: 10-12-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 70374-39-9
Molecular Formula C13 H10 Cl N3 O4 S2
Molecular Weight (g/mol) 371.82
Synonym 6-Chloro-4-hydroxy-2-methyl-N-2-pyridinyl-2H-thieno[2,3-e]-1,2-thiazine-3-carboxamide 1,1-dioxide, Chlortenoxicam, Fulactive, Lofecam, Lorcam, Lornoxicam, Ro 13-9297, TS 110, Telos, Xefo, Xefocam
IUPAC Name 6-chloro-4-hydroxy-2-methyl-1,1-dioxo-N-pyridin-2-ylthieno[2,3-e]thiazine-3-carboxamide
SMILES CN1C(=C(O)c2sc(Cl)cc2S1(=O)=O)C(=O)Nc3ccccn3

Specifications

Quantity 10 mg
InChI Formula InChI=1S/C13H10ClN3O4S2/c1-17-10(13(19)16-9-4-2-3-5-15-9)11(18)12-7(23(17,20)21)6-8(14)22-12/h2-6,18H,1H3,(H,15,16,19)
Formula Weight 370.9801
Percent Purity >95%
Chemical Name or Material Lornoxicam
Grade HPLC
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.