missing translation for 'onlineSavingsMsg'
Learn More

Lornoxicam-d4, TRC

Product Code. 30466709
Change view
Click to view available options
Quantity:
5 mg
Unit Size:
5mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30466709 5 mg 5mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Product Code. 30466709 Supplier Toronto Research Chemicals Supplier No. TRCL4700025MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Lornoxicam-d4
CAS 1216527-48-8
Molecular Formula C13 2H4 H6 Cl N3 O4 S2
Quantity 5 mg
Synonym 6-Chloro-4-hydroxy-2-methyl-1,1-dioxo-N-(3,4,5,6-tetradeuteriopyridin-2-yl)thieno[2,3-e]thiazine-3-carboxamide, 6-Chloro-4-hydroxy-2-methyl-N-(2-pyridinyl-3,4,5,6-d4)-2H-thieno[2,3-e]-1,2-thiazine-3-carboxamide 1,1-dioxide, Chlortenoxicam-d4, Lornoxicam-d4, Lornoxicam-D4 (pyridyl-D4), Lornoxicam-d4 (pyridine-d4)
InChI Formula InChI=1S/C13H10ClN3O4S2/c1-17-10(13(19)16-9-4-2-3-5-15-9)11(18)12-7(23(17,20)21)6-8(14)22-12/h2-6,18H,1H3,(H,15,16,19)/i2D,3D,4D,5D
SMILES [2H]c1nc(NC(=O)C2=C(O)c3sc(Cl)cc3S(=O)(=O)N2C)c([2H])c([2H])c1[2H]
IUPAC Name 6-chloro-4-hydroxy-2-methyl-1,1-dioxo-N-(3,4,5,6-tetradeuteriopyridin-2-yl)thieno[2,3-e]thiazine-3-carboxamide
Molecular Weight (g/mol) 375.84
Formula Weight 375.0052
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.