missing translation for 'onlineSavingsMsg'
Learn More

Lorglumide Sodium Salt, TRC

Product Code. 30469434
Change view
Click to view available options
Quantity:
50 mg
Unit Size:
50mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30469434 50 mg 50mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30469434 Supplier Toronto Research Chemicals Supplier No. TRCL46991050MG
 Restricted Item
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Lorglumide Sodium Salt
CAS 1021868-76-7
Molecular Formula C22H31Cl2N2NaO4
Quantity 50 mg
Synonym 4-[(3,4-Dichlorobenzoyl)amino]-5-(dipentylamino)-5-oxo-pentanoic Acid Sodium Salt
InChI Formula InChI=1S/C22H32Cl2N2O4.Na/c1-3-5-7-13-26(14-8-6-4-2)22(30)19(11-12-20(27)28)25-21(29)16-9-10-17(23)18(24)15-16;/h9-10,15,19H,3-8,11-14H2,1-2H3,(H,25,29)(H,27,28);/q;+1/p-1
SMILES O=C(O[Na])CCC(NC(C1=CC=C(Cl)C(Cl)=C1)=O)C(N(CCCCC)CCCCC)=O
IUPAC Name sodium;4-[(3,4-dichlorobenzoyl)amino]-5-(dipentylamino)-5-oxopentanoate
Molecular Weight (g/mol) 481.39
Formula Weight 480.1559
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.