missing translation for 'onlineSavingsMsg'
Learn More

Lorglumide sodium salt, MedChemExpress

Product Code. 16421118
Encompass_Preferred
Change view
Click to view available options
Quantity:
5 mg
10 mg
Unit Size:
10mg
5mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
16421118 5 mg 5mg
16408708 10 mg 10mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 16421118 Supplier Medchem Express Supplier No. HYB1439B
 Restricted Item
Estimated Shipment: 16-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

MedChemExpress Lorglumide sodium salt (CR-1409 sodium salt) is a potent cholecystokinin (CCK) receptor antagonist.

Lorglumide sodium salt (CR-1409 sodium salt) is a potent cholecystokinin (CCK) receptor antagonist.

Specifications

Chemical Name or Material Lorglumide sodium salt
CAS 1021868-76-7
Color White
Purity Grade Notes Research
Molecular Formula C22H31Cl2N2NaO4
Quantity 5 mg
Synonym CR-1409 sodium salt
Solubility Information H2O : ≥ 100 mg/mL (207.73 mM) ∣DMSO : 100 mg/mL (207.73 mM; Need ultrasonic)
SMILES ClC1=C(C=CC(C(NC(CCC(O[Na])=O)C(N(CCCCC)CCCCC)=O)=O)=C1)Cl
Molecular Weight (g/mol) 481.39
Formula Weight 481.39
Percent Purity 99.0%
Grade Research
Physical Form Powder
For Use With (Application) Metabolism-protein/nucleotide metabolism
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.