missing translation for 'onlineSavingsMsg'
Learn More

Loreclezole, MedChemExpress

Product Code. 16466737
Encompass_Preferred
Change view
Click to view available options
Quantity:
5 mg
10 mg
50 mg
100 mg
Unit Size:
100mg
10mg
50mg
5mg
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16466737 5 mg 5mg
16445016 100 mg 100mg
16444028 10 mg 10mg
16400387 50 mg 50mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
This item is not returnable. View return policy
Product Code. 16466737 Supplier Medchem Express Supplier No. HY105272

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress Loreclezole, an antiepileptic compound, is a selective GABAA receptor modulator and acts as a positive allosteric modulator of β2 or β3-subunit containing receptors.

Loreclezole, an antiepileptic compound, is a selective GABAA receptor modulator and acts as a positive allosteric modulator of β2 or β3-subunit containing receptors.

Specifications

Chemical Name or Material Loreclezole
Physical Form Solid
CAS 117857-45-1
Color White
Quantity 5 mg
Purity Grade Notes Research
Molecular Formula C10H6Cl3N3
For Use With (Application) Neuroscience-Neuromodulation
Synonym R 72063
Solubility Information DMSO : 110 mg/mL (400.68 mM; Need ultrasonic) ∣H2O : < 0.1 mg/mL (insoluble)
SMILES Cl/C(C1=C(C=C(Cl)C=C1)Cl)=C\N2C=NC=N2
Molecular Weight (g/mol) 274.53
Formula Weight 274.53
Percent Purity 99.68%
Grade Research
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.