missing translation for 'onlineSavingsMsg'
Learn More

Lorcaserin, TRC

Product Code. 17943925
Change view
Click to view available options
Quantity:
2.5 mg
5 mg
Unit Size:
2.50mg
5mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
17943925 5 mg 5mg
17943995 2.5 mg 2.50mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 17943925 Supplier Toronto Research Chemicals Supplier No. TRCL2158555MG
 Restricted Item

This item requires a price update prior to being ordered.

Request a Price for this item here.

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 616202-92-7
Molecular Formula C11H14ClN
Molecular Weight (g/mol) 195.69
Synonym 1H-3-Benzazepine, 8-chloro-2,3,4,5-tetrahydro-1-methyl-, (1R)-, (1R)-8-Chloro-2,3,4,5-tetrahydro-1-methyl-1H-3-benzazepine, (R)-8-Chloro-1-methyl-2,3,4,5-tetrahydro-1H-3-benzazepine, Belviq, Lorcaserin
IUPAC Name (5R)-7-chloro-5-methyl-2,3,4,5-tetrahydro-1H-3-benzazepine
SMILES C[C@H]1CNCCc2ccc(Cl)cc12

Specifications

Quantity 5 mg
InChI Formula InChI=1S/C11H14ClN/c1-8-7-13-5-4-9-2-3-10(12)6-11(8)9/h2-3,6,8,13H,4-5,7H2,1H3/t8-/m0/s1
Formula Weight 195.0815
Percent Purity >95%
Chemical Name or Material Lorcaserin
Grade HPLC
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.