missing translation for 'onlineSavingsMsg'
Learn More

Loratadine, MedChemExpress

Product Code. 16467246
Encompass_Preferred
Change view
Click to view available options
Quantity:
10 mg
50 mg
100 mg
Unit Size:
100mg
10mg
50mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
16467246 100 mg 100mg
16477288 10 mg 10mg
16443307 50 mg 50mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 16467246 Supplier Medchem Express Supplier No. HY17043
 Restricted Item
Estimated Shipment: 14-12-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

MedChemExpress Loratadine (SCH-29851) is a selective inverse peripheral histamine H1-receptor agonist with an IC50 of >32 μM. Loratadine has anti-dengue-virus (DENV) activity. Loratadine can inhibit immunologic release of inflammatory mediators.

Loratadine (SCH-29851) is a selective inverse peripheral histamine H1-receptor agonist with an IC50 of >32 μM. Loratadine has anti-dengue-virus (DENV) activity. Loratadine can inhibit immunologic release of inflammatory mediators.

Specifications

Chemical Name or Material Loratadine
Physical Form Powder
CAS 79794-75-5
Color White
Quantity 100 mg
Purity Grade Notes Research
Molecular Formula C22H23ClN2O2
For Use With (Application) COVID-19-anti-virus
Synonym Loratidine SCH 29851
Solubility Information DMSO : 50 mg/mL (130.59 mM; Need ultrasonic)
SMILES O=C(N1CC/C(CC1)=C2C3=CC=C(Cl)C=C3CCC4=CC=CN=C4\2)OCC
Molecular Weight (g/mol) 382.88
Formula Weight 382.88
Percent Purity 99.6%
Grade Research
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.