missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Lopinavir, LoGiCal
Click to view available options
Quantity:
10 mg
Unit Size:
10mg
Specifications
Specifications
| Chemical Name or Material | Lopinavir |
| CAS | 192725-17-0 |
| Analyte or Component Names | Lopinavir |
| Shipping Condition | Room Temperature |
| Molecular Formula | C37 H48 N4 O5 |
| Certifications/Compliance | ISO 9001 |
| Synonym | Lopinavir, (2S)-N-[(1S,3S,4S)-1-Benzyl-4-[[2-(2,6-dimethylphenoxy)acetyl]amino]-3-hydroxy-5-phenylpentyl]-3-methyl-2-[2-oxotetrahydropyrimidin-1(2H)-yl]butanamide, 1(2H)-Pyrimidineacetamide, N-[(1S,3S,4S)-4-[[2-(2,6-dimethylphenoxy)acetyl]amino]-3-hydroxy-5-phenyl-1-(phenylmethyl)pentyl]tetrahydro-alpha-(1-methylethyl)-2-oxo-, (alphaS)-, 1(2H)-Pyrimidineacetamide, N-[(1S,3S,4S)-4-[[(2,6-dimethylphenoxy)acetyl]amino]-3-hydroxy-5-phenyl-1-(phenylmethyl)pentyl]tetrahydro-alpha-(1-methylethyl)-2-oxo-, (alphaS)- (9CI), 1(2H)-Pyrimidineacetamide, N-[4-[[(2,6-dimethylphenoxy)acetyl]amino]-3-hydroxy-5-phenyl-1-(phenylmethyl)pentyl]tetrahydro-alpha-(1-methylethyl)-2-oxo-, [1S-[1R*(R*),3R*,4R*]]-, (alphaS)-N-[(1S,3S,4S)-4-[[2-(2,6-Dimethylphenoxy)acetyl]amino]-3-hydroxy-5-phenyl-1-(phenylmethyl)pentyl]tetrahydro-alpha-(1-methylethyl)-2-oxo-1(2H)-pyrimidineacetamide, A 157378.0, ABT 378, Aluviran, Koletra |
| InChI Formula | InChI=1S/C37H48N4O5/c1-25(2)34(41-20-12-19-38-37(41)45)36(44)39-30(21-28-15-7-5-8-16-28)23-32(42)31(22-29-17-9-6-10-18-29)40-33(43)24-46-35-26(3)13-11-14-27(35)4/h5-11,13-18,25,30-32,34,42H,12,19-24H2,1-4H3,(H,38,45)(H,39,44)(H,40,43)/t30-,31-,32-,34-/m0/s1 |
| Solution Type | Neat |
| SMILES | CC(C)[C@H](N1CCCNC1=O)C(=O)N[C@H](C[C@H](O)[C@H](Cc2ccccc2)NC(=O)COc3c(C)cccc3C)Cc4ccccc4 |
| Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction