missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Tocris Bioscience™ Lopinavir

Click to view available options
Quantity:
50 mg
Unit Size:
50mg
Description
Lopinavir is a highly potent and selective HIV-1 protease inhibitor (IC50 = 6.5 nM, HIV cultured in peripheral blood mononuclear cells; Ki = 1.3 pM). Displays >105-fold selectivity for HIV-1 protease over mammalian proteases including renin and cathepsin D and E. Inhibits HIV-1 protease in patient HIV isolates displaying mutations that confer resistance to ritonavir (Cat. No. 5856) (IC50 values in 5 - 50 nM range). Also inhibits SARS-CoV-2 replication in vitro (IC50 = 26 μM)
Specifications
Specifications
| Chemical Name or Material | (αS)-N-[(1 S,3 S,4 S)-4-[[2-(2,6-Dimethylphenoxy)acetyl]amino]-3-hydroxy-5-phenyl-1-(phenylmethyl)pentyl]tetrahydro-α-(1-methylethyl)-2-oxo-1(2 H)-pyrimidineacetamide |
| CAS | 192725-17-0 |
| Quantity | 50 mg |
| Target | Additional Synthetic Protease Inhibitors |
| Molecular Formula | C37H48N4O5 |
| Purity | 0.98 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction