missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Loperamide hydrochloride, Tocris Bioscience™
Description
High affinity μ-opioid receptor agonist with peripheral selectivity (Ki values are 2, 48 and 1156 nM for μ-, δ- and κ-opioid receptors respectively). Antidiarrhoeal and antihyperalgesic agent. Also a Ca2+ channel blocker; at low micromolar concentrations it blocks broad spectrum neuronal HVA Ca2+ channels and at higher concentrations it reduces Ca2+ flux through NMDA receptor operated channels.
Specifications
Specifications
| CAS | 34552-83-5 |
| Molecular Formula | C29H34Cl2N2O2 |
| Quantity | 1 g |
| Synonym | loperamide hydrochloride, loperamide hcl, imodium, suprasec, dissenten, fortasec, lopemid, lopemin, loperyl, imosec |
| InChI Key | PGYPOBZJRVSMDS-UHFFFAOYSA-N |
| SMILES | CN(C)C(=O)C(CCN1CCC(CC1)(C2=CC=C(C=C2)Cl)O)(C3=CC=CC=C3)C4=CC=CC=C4.Cl |
| IUPAC Name | 4-[4-(4-chlorophenyl)-4-hydroxypiperidin-1-yl]-N,N-dimethyl-2,2-diphenylbutanamide;hydrochloride |
| Molecular Weight (g/mol) | 513.503 |
| PubChem CID | 71420 |
| ChEBI | CHEBI:6533 |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?