missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Loperamide hydrochloride, MedChemExpress
Click to view available options
Quantity:
100 mg
500 mg
Unit Size:
100mg
500mg
Description
Loperamide (hydrochloride) (R-18553 (hydrochloride)) is an opioid receptor agonist. Loperamide hydrochloride is a selective and competitive human intestinal carboxylesterases (hiCE) inhibitor. Loperamide hydrochloride has anti-diarrheal effect.
Specifications
Specifications
| Chemical Name or Material | Loperamide hydrochloride |
| CAS | 34552-83-5 |
| Color | White |
| Purity Grade Notes | Research |
| Molecular Formula | C29H34Cl2N2O2 |
| Quantity | 500 mg |
| Synonym | R-18553 hydrochloride |
| Solubility Information | DMSO : 50 mg/mL (97.37 mM; Need ultrasonic) ∣H2O : 1 mg/mL (1.95 mM; Need ultrasonic) |
| SMILES | O=C(N(C)C)C(C1=CC=CC=C1)(C2=CC=CC=C2)CCN3CCC(O)(C4=CC=C(Cl)C=C4)CC3.Cl |
| Molecular Weight (g/mol) | 513.5 |
| Show More |
Research purposes only
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction