missing translation for 'onlineSavingsMsg'
Learn More

Loperamide hydrochloride, MedChemExpress

Product Code. 16428296
Encompass_Preferred
Click to view available options
Quantity:
100 mg
500 mg
Unit Size:
100mg
500mg
This item is not returnable. View return policy

Product Code. 16428296

Brand: Medchem Express HYB0418A

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress Loperamide (hydrochloride) (R-18553 (hydrochloride)) is an opioid receptor agonist. Loperamide hydrochloride is a selective and competitive human intestinal carboxylesterases (hiCE) inhibitor. Loperamide hydrochloride has anti-diarrheal effect.

Loperamide (hydrochloride) (R-18553 (hydrochloride)) is an opioid receptor agonist. Loperamide hydrochloride is a selective and competitive human intestinal carboxylesterases (hiCE) inhibitor. Loperamide hydrochloride has anti-diarrheal effect.

Specifications

Chemical Name or Material Loperamide hydrochloride
CAS 34552-83-5
Color White
Purity Grade Notes Research
Molecular Formula C29H34Cl2N2O2
Quantity 100 mg
Synonym R-18553 hydrochloride
Solubility Information DMSO : 50 mg/mL (97.37 mM; Need ultrasonic) ∣H2O : 1 mg/mL (1.95 mM; Need ultrasonic)
SMILES O=C(N(C)C)C(C1=CC=CC=C1)(C2=CC=CC=C2)CCN3CCC(O)(C4=CC=C(Cl)C=C4)CC3.Cl
Molecular Weight (g/mol) 513.5
Formula Weight 513.5
Percent Purity 99.79%
Grade Research
Physical Form Solid
For Use With (Application) Metabolism-protein/nucleotide metabolism
Show More Show Less

Research purposes only

Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.