missing translation for 'onlineSavingsMsg'
Learn More
Learn More
SeaPlaque™ Agarose, Lonza™
Click to view available options
Quantity:
25 g
125 g
Unit Size:
125g
Each
Description
- For preparative DNA and RNA electrophoresis
- Molecular biology grade
- ≤0.10 EEO
- Low melting temperature argarose (26°–30°C Gelling temperature)
- DNase/RNase-free
Specifications
Specifications
| Chemical Name or Material | SeaPlaque™ Agarose |
| Quantity | 25 g |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction