missing translation for 'onlineSavingsMsg'
Learn More

SeaKem™ ME Agarose, Lonza™

Product Code. 11507775
Click to view available options
Quantity:
25 g
125 g
500 g
Unit Size:
125g
25g
Each
This item is not returnable. View return policy

Product Code. 11507775

Brand: Lonza 50014

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

  • For serum protein electrophoresis and nucleic acid electrophoresis

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material SeaKem ME Agarose
Quantity 500 g
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.