missing translation for 'onlineSavingsMsg'
Learn More

Lonza™ ProSieve™ Color Protein Markers

Product Code. 11547995
Change view
Click to view available options
Quantity:
100 μL
500 μL
Range:
10 to 190 kd
Unit Size:
100µL
500µL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Range unitSize
11547995 500 μL 10 to 190 kd 500µL
11557995 100 μL 10 to 190 kd 100µL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11547995 Supplier Lonza™ Supplier No. 50550

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Visible markers for use in SDS-PAGE gels

  • Useful for monitoring the efficiency of separation during electrophoresis
  • Confirm that transfer has occurred from gel to membrane in Western blotting

ProSieve Color Protein Marker

  • Proteins in the marker have been labeled with 3 different fluorescent dyes
  • Marker is supplied as lyophilized solid, add water, warm briefly at 37°C and load.
  • Proteins have approximate masses of 10, 15, 20, 25, 40, 50, 80, 125, and 190kDa
  • Each lot is individually calibrated against a ProSieve Unstained Protein Marker
    
ProSieve QuadColor Protein Marker
  • Proteins are individually prestained using four different dyes, producing a brightly colored ladder with an easy-to-remember pattern
  • Marker is supplied ready-to-use, no heating, further dilution or addition of a reducing agent is required before use
  • Mixture of 12 recombinant, highly purified proteins with molecular weights of approximately 4, 6, 10, 15, 25, 40, 55, 70, 100, 140, 170, 250, and 300kDa
  • Apparent molecular weights are provided for each lot

TRUSTED_SUSTAINABILITY

Specifications

Form Liquid
Formulation Markers have been labeled with 3 different Fluorescent dyes, Marker is supplied as lyophilized solid, add water, warm briefly at 37°C and load
For Use With (Application) Monitoring protein separation prior to staining and provide confirmation of protein transfer in Western blotting
Quantity 500 μL
Range 10 to 190 kd
Storage Requirements −20°C
Product Type Protein Molecular Weight Marker
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.