missing translation for 'onlineSavingsMsg'
Learn More

NuSieve™ 3:1 Agarose, Lonza™

Product Code. p-3148404
Click to view available options
Quantity:
25 g
500 g
Unit Size:
Each
This item is not returnable. View return policy

Product Code. 11537785

Brand: Lonza 50091

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Easy-to-handle gels enhance the speed of gel processing and blotting. Lonza™ NuSieve™ 3:1 Agarose is a molecular biology grade, standard melting temperature agarose that yields strong gels for fine resolution of small DNA, RNA, and PCR products ≤ 1 kb.

  • Designed for analytical electrophoresis
  • Resolution of DNA fragments up to 1,000 bp, but is capable of resolving DNA fragments  from 10 bp up to 1,500 bp
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 9012-36-6

Specifications

Quantity 25 g
Grade Molecular Biology
Chemical Name or Material Agarose
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.