missing translation for 'onlineSavingsMsg'
Learn More
Learn More
NuSieve™ 3:1 Agarose, Lonza™
Click to view available options
Quantity:
25 g
500 g
Unit Size:
Each
Description
- Designed for analytical electrophoresis
- Resolution of DNA fragments up to 1,000 bp, but is capable of resolving DNA fragments from 10 bp up to 1,500 bp
Specifications
Specifications
| Quantity | 500 g |
| Grade | Molecular Biology |
| Chemical Name or Material | Agarose |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction